Gene Transcripts

Gene Symbol
Prss39
Entrez Gene ID
363215
Gene Name
protease, serine, 39
Gene Aliases
Prss40, Tesp1, Tesp2
Chromosome Location
Chr. 9: 41006606 - 41012607 on Build Rnor_6.0
UniGene ID
Rn.140412
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013226.1 NP_001013244.1 2, 3 636
XM_006244717.2 XP_006244779.1 3, 4 692
XM_006244718.2 XP_006244780.1 3, 4 320
GenBank mRNA BC079254.1 - 2, 3 636

View Full Product Details