Gene Transcripts

Gene Symbol
Atg9a
Entrez Gene ID
363254
Gene Name
autophagy related 9A
Gene Aliases
RGD1310450
Chromosome Location
Chr. 9: 82382800 - 82393428 on Build Rnor_6.0
UniGene ID
Rn.35248
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014218.1 NP_001014240.1 6 824
XM_006245247.2 XP_006245309.1 6 528
XM_008767262.1 XP_008765484.1 6 1191
XM_008767263.1 XP_008765485.1 6 703
XM_008767264.1 XP_008765486.1 6 531
XM_008767265.1 XP_008765487.1 7 589
XM_008767266.1 XP_008765488.1 7 592
GenBank mRNA BC089204.1 - 6 824

View Full Product Details