Gene Transcripts

Gene Symbol
Ift46
Entrez Gene ID
300675
Gene Name
intraflagellar transport 46
Gene Aliases
RGD1307682
Chromosome Location
Chr. 8: 49075954 - 49097689 on Build Rnor_6.0
UniGene ID
Rn.13688
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024760.1 NP_001019931.1 6 623
XM_006242930.2 XP_006242992.1 7 762
XM_006242931.2 XP_006242993.1 6 903
GenBank mRNA BC079388.1 - 6 529
BC097954.1 - 6 623 7456463

View Full Product Details