Gene Transcripts

Gene Symbol
Klk1b22
Entrez Gene ID
13646
Gene Name
kallikrein 1-related peptidase b22
Gene Aliases
Egfbp-1, Egfbp1, Klk22, mGk-22
Chromosome Location
Chr. 7: 44112683 - 44116876 on Build GRCm38
UniGene ID
Mm.5193
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010114.1 NP_034244.1 5 721
GenBank mRNA BC106155.1 - 5 725

View Full Product Details