Gene Transcripts

Gene Symbol
Dusp23
Entrez Gene ID
68440
Gene Name
dual specificity phosphatase 23
Gene Aliases
1300005N15Rik, LDP-3
Chromosome Location
Chr. 1: 172630769 - 172632974 on Build GRCm38
UniGene ID
Mm.33024
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026725.2 NP_081001.1 2 538
GenBank mRNA AB183013.1 - 2 532
AK004912.1 - 2 429
BC069187.1 - 2 532 850475

View Full Product Details