Gene Transcripts
- Gene Symbol
- LYAR
- Entrez Gene ID
- 55646
- Gene Name
- Ly1 antibody reactive
- Gene Aliases
- ZC2HC2, ZLYAR
- Chromosome Location
- Chr. 4: 4267701 - 4290169 on Build GRCh38
- UniGene ID
- Hs.425427
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145725.1 |
NP_001139197.1
|
9, 10 |
1149 |
|
NM_017816.2 |
NP_060286.1
|
9, 10 |
1247 |
|
XM_011513505.1 |
XP_011511807.1
|
9, 10 |
1549 |
|
XM_011513506.2 |
XP_011511808.1
|
8, 9 |
1180 |
|
GenBank mRNA |
AF229835.1 |
- |
3, 4 |
200 |
|
AK000432.1 |
- |
9, 10 |
1194 |
|
AK290750.1 |
- |
9, 10 |
1224 |
|
AL136750.1 |
- |
9, 10 |
1149 |
|
BC015796.2 |
- |
9, 10 |
1209 |
3911275 |
CR533548.1 |
- |
7, 8 |
1006 |
|
View Full Product Details