Gene Transcripts

Gene Symbol
SPRN
Entrez Gene ID
503542
Gene Name
shadow of prion protein homolog (zebrafish)
Gene Aliases
SHADOO, SHO, bA108K14.1
Chromosome Location
Chr. 10: 133420666 - 133424617 on Build GRCh38
UniGene ID
Hs.64968
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012508.3 NP_001012526.2 2 1308
GenBank mRNA BC040198.1 - 2 1268
CD580523.2 - 1 602

View Full Product Details