Gene Transcripts

Gene Symbol
Itpripl1
Entrez Gene ID
499885
Gene Name
inositol 1,4,5-trisphosphate receptor interacting protein-like 1
Gene Aliases
Chromosome Location
Chr. 3: 119616473 - 119619865 on Build Rnor_6.0
UniGene ID
Rn.123061
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025043.1 NP_001020214.1 2 1541
XM_006234927.2 XP_006234989.1 2 2054
GenBank mRNA BC082013.1 - 2 1541

View Full Product Details