Gene Transcripts

Gene Symbol
THAP3
Entrez Gene ID
90326
Gene Name
THAP domain containing 3
Gene Aliases
Chromosome Location
Chr. 1: 6624645 - 6635586 on Build GRCh38
UniGene ID
Hs.689006
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138350.3 NP_612359.2 4, 5 521
XM_017002761.1 XP_016858250.1 4, 5 458
GenBank mRNA AK096217.1 - 4, 5 479

View Full Product Details