Gene Transcripts

Gene Symbol
Yipf1
Entrez Gene ID
298312
Gene Name
Yip1 domain family, member 1
Gene Aliases
Chromosome Location
Chr. 5: 126932738 - 126968110 on Build Rnor_6.0
UniGene ID
Rn.98698
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199383.2 NP_955415.1 10 1220
GenBank mRNA BC062239.1 - 10 1224
FQ218908.1 - 10 1219

View Full Product Details