Gene Transcripts

Gene Symbol
CLN6
Entrez Gene ID
54982
Gene Name
ceroid-lipofuscinosis, neuronal 6, late infantile, variant
Gene Aliases
CLN4A, HsT18960, nclf
Chromosome Location
Chr. 15: 68206992 - 68229742 on Build GRCh38
UniGene ID
Hs.584921
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017882.2 NP_060352.1 7 1343
GenBank mRNA AK000568.1 - 7 1284
AK027604.1 - 7 1307
AK074753.1 - 4 833
AK291175.1 - 7 1266
BC010849.1 - 7 1253 3878776
BC013130.1 - 7 1190 4517748
CN427564.1 - 1 109

View Full Product Details