Gene Transcripts

Gene Symbol
PHIP
Entrez Gene ID
55023
Gene Name
pleckstrin homology domain interacting protein
Gene Aliases
BRWD2, DCAF14, WDR11, ndrp
Chromosome Location
Chr. 6: 78934419 - 79078294 on Build GRCh38
UniGene ID
Hs.511817
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017934.5 NP_060404.3 30 3745
XM_005248729.4 XP_005248786.1 30 3742
XM_011535918.2 XP_011534220.1 27 3401
XM_017010989.1 XP_016866478.1 32 3887
XM_017010990.1 XP_016866479.1 29 3485
GenBank mRNA AF310250.1 - 7 752
AJ303102.1 - 30 3532
AK000712.1 - 3 375
AL161957.1 - 8 808
BC008909.2 - 5 473
BC064611.1 - 2 214
BC081569.1 - 8 788
BC137488.1 - 30 3625 9021488
DQ924532.1 - 30 3519

View Full Product Details