Gene Transcripts

Gene Symbol
Rnaseh2b
Entrez Gene ID
67153
Gene Name
ribonuclease H2, subunit B
Gene Aliases
1110019N06Rik, 2610207P08Rik, Dleu8
Chromosome Location
Chr. 14: 62332068 - 62392708 on Build GRCm38
UniGene ID
Mm.26040
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026001.2 NP_080277.1 5 381
GenBank mRNA AK011914.1 - 5 420
AK082752.1 - 5 382
AK163912.1 - 2 3227
BC047997.1 - 5 382 6510494

View Full Product Details