Gene Transcripts

Gene Symbol
Vps39
Entrez Gene ID
269338
Gene Name
vacuolar protein sorting 39 (yeast)
Gene Aliases
A230065P22Rik, AW743070, Vam6, Vam6P, mVam6
Chromosome Location
Chr. 2: 120316461 - 120353144 on Build GRCm38
UniGene ID
Mm.246131
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147153.3 NP_671495.1 4 336
NM_178851.3 NP_849182.1 3 303
GenBank mRNA AB028844.1 - 3 283
AF281050.1 - 4 313
AF281051.1 - 3 280
AK038824.1 - 3 292
AK042253.1 - 3 292
AK084743.1 - 3 303
AK129210.1 - 3 298
AK140041.1 - 4 341
AK141424.1 - 3 286
AK154103.1 - 3 292
AK159889.1 - 3 296
AK160109.1 - 3 296
AK170831.1 - 4 363
AK171008.1 - 3 292
BB655861.1 - 4 329
BC007479.1 - 3 254 3492875
BY125125.1 - 3 304

View Full Product Details