Gene Transcripts

Gene Symbol
Rxfp3
Entrez Gene ID
294807
Gene Name
relaxin/insulin-like family peptide receptor 3
Gene Aliases
GPCR135, RGD1311550, Rln3r1, Salpr
Chromosome Location
Chr. 2: 61006337 - 61007767 on Build Rnor_6.0
UniGene ID
Rn.127658
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008310.1 NP_001008311.1 1 1015
GenBank mRNA AY633763.1 - 1 994
AY633764.1 - 1 1015

View Full Product Details