Gene Transcripts

Gene Symbol
CUEDC1
Entrez Gene ID
404093
Gene Name
CUE domain containing 1
Gene Aliases
Chromosome Location
Chr. 17: 57861243 - 57955323 on Build GRCh38
UniGene ID
Hs.46679
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271875.1 NP_001258804.1 8 1372
NM_001292025.1 NP_001278954.1 8 1677
XM_006721900.2 XP_006721963.1 8 1987
XM_011524812.2 XP_011523114.1 8 760
XM_017024640.1 XP_016880129.1 7 1258
XM_017024641.1 XP_016880130.1 9 1389
GenBank mRNA AK000746.1 - 8 1677
AK000977.1 - 7 1027
BC056882.1 - 8 1372 6150503
CR627470.1 - 3 170

View Full Product Details