Gene Transcripts

Gene Symbol
Lce3f
Entrez Gene ID
69520
Gene Name
late cornified envelope 3F
Gene Aliases
2310002A05Rik
Chromosome Location
Chr. 3: 92992226 - 92993426 on Build GRCm38
UniGene ID
Mm.439968
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018079.1 NP_001018089.1 2 412
GenBank mRNA AK009081.1 - 1 264
BC060281.1 - 2 412 30288382

View Full Product Details