Gene Transcripts

Gene Symbol
Chac2
Entrez Gene ID
360994
Gene Name
ChaC, cation transport regulator homolog 2 (E. coli)
Gene Aliases
RGD1309120
Chromosome Location
Chr. 14: 115383906 - 115391769 on Build Rnor_6.0
UniGene ID
Rn.234312
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025016.1 NP_001020187.1 3 926
GenBank mRNA BC082031.1 - 3 926

View Full Product Details