Gene Transcripts
- Gene Symbol
- Sry
- Entrez Gene ID
- 25221
- Gene Name
- sex determining region Y
- Gene Aliases
- SRYGENE, Sry1, Sry3BI, Tdf, Tdy
- Chromosome Location
- Chr. Y: 327176 - 327685 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012772.1 |
NP_036904.1
|
1 |
233 |
|
- Gene Symbol
- LOC103694552
- Entrez Gene ID
- 103694552
- Gene Name
- sex-determining region Y protein
- Gene Aliases
- Chromosome Location
- Chr. Y: 548637 - 551544 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details