Gene Transcripts

Gene Symbol
ANO1
Entrez Gene ID
55107
Gene Name
anoctamin 1
Gene Aliases
DOG1, ORAOV2, TAOS2, TMEM16A
Chromosome Location
Chr. 11: 70078302 - 70189545 on Build GRCh38
UniGene ID
Hs.503074
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018043.5 NP_060513.5 13, 14 1666
XM_006718602.2 XP_006718665.1 14, 15 1448
XM_006718604.2 XP_006718667.1 14, 15 1563
XM_011545121.2 XP_011543423.1 14, 15 1440
XM_011545123.2 XP_011543425.1 14, 15 1437
XM_011545125.2 XP_011543427.1 14, 15 1440
XM_011545127.2 XP_011543429.1 13, 14 1374
XM_011545129.2 XP_011543431.1 14, 15 1355
XM_017017956.1 XP_016873445.1 13, 14 1371
GenBank mRNA AB845669.1 - 13, 14 1361
AK001123.1 - 4, 5 282
AK293088.1 - 13, 14 1374
AL833582.1 - 13, 14 1410
AY728143.1 - 13, 14 1666
BC027590.2 - 8, 9 618

View Full Product Details