Gene Transcripts

Gene Symbol
Ffar1
Entrez Gene ID
233081
Gene Name
free fatty acid receptor 1
Gene Aliases
Gpr40
Chromosome Location
Chr. 7: 30860568 - 30861470 on Build GRCm38
UniGene ID
Mm.425422
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194057.2 NP_918946.2 1 811
GenBank mRNA AB095745.1 - 1 811
BC132014.1 - 1 905 40130291
BC138099.1 - 1 905 8861118

View Full Product Details