Gene Transcripts

Gene Symbol
Bri3bp
Entrez Gene ID
498176
Gene Name
Bri3 binding protein
Gene Aliases
Chromosome Location
Chr. 12: 36576018 - 36587839 on Build Rnor_6.0
UniGene ID
Rn.136726
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017487.1 NP_001017487.1 3 718
GenBank mRNA BC085334.1 - 3 718

View Full Product Details