Gene Transcripts

Gene Symbol
Trappc8
Entrez Gene ID
75964
Gene Name
trafficking protein particle complex 8
Gene Aliases
5033403J15Rik, AI845423, D030074E01Rik, Trs85, mKIAA1012
Chromosome Location
Chr. 18: 20817223 - 20896386 on Build GRCm38
UniGene ID
Mm.273769
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029491.2 NP_083767.2 7 994
NM_177038.3 NP_796012.2 7 994
GenBank mRNA AK083747.1 - 7 994

View Full Product Details