Gene Transcripts

Gene Symbol
Bri3bp
Entrez Gene ID
76809
Gene Name
Bri3 binding protein
Gene Aliases
2410150I18Rik, AI841257, AW742481
Chromosome Location
Chr. 5: 125441538 - 125460885 on Build GRCm38
UniGene ID
Mm.290720
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029752.2 NP_084028.1 3 1031
GenBank mRNA AK043288.1 - 3 973
AK044565.1 - 3 919
AK143538.1 - 3 990

View Full Product Details