Gene Transcripts

Gene Symbol
Krt31
Entrez Gene ID
450228
Gene Name
keratin 31, type I
Gene Aliases
Ka25
Chromosome Location
Chr. 10: 87959123 - 87962891 on Build Rnor_6.0
UniGene ID
Rn.129815
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008817.1 NP_001008817.1 7 1141
XM_006247392.2 XP_006247454.1 7 1192

View Full Product Details