Gene Transcripts

Gene Symbol
Adam26a
Entrez Gene ID
13525
Gene Name
a disintegrin and metallopeptidase domain 26A (testase 3)
Gene Aliases
Adam26, Dtgn4
Chromosome Location
Chr. 8: 43568273 - 43576707 on Build GRCm38
UniGene ID
Mm.334510
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010085.2 NP_034215.2 2 2198
GenBank mRNA AF167404.1 - 2 2157
AK029756.1 - 2 2199

View Full Product Details