Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016956.3 | NP_058652.1 | 2 | 281 | |
GenBank mRNA | AB020014.1 | - | 2 | 224 | |
AB020016.1 | - | 2 | 224 | ||
AB020017.1 | - | 2 | 224 | ||
AB364478.1 | - | 2 | 227 | ||
AF149782.1 | - | 2 | 227 | ||
BC027434.1 | - | 2 | 261 | 4920093 | |
BC032264.1 | - | 2 | 272 | 3988455 |