Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163282.2 | NP_001156754.1 | 3 | 662 | |
GenBank mRNA | AK162823.1 | - | 3 | 645 | |
BC094351.1 | - | 3 | 610 | ||
M62301.1 | - | 3 | 650 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138647.3 | NP_619588.1 | 3 | 662 | |
GenBank mRNA | AK053885.1 | - | 3 | 665 | |
BC132318.1 | - | 3 | 607 | 40130595 |