Gene Transcripts

Gene Symbol
LOC202181
Entrez Gene ID
202181
Gene Name
SUMO interacting motifs containing 1 pseudogene
Gene Aliases
Chromosome Location
Chr. 5: 177618500 - 177672277 on Build GRCh38
UniGene ID
Hs.189914
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK297457.1 - 1, 2 261

Gene Symbol
SIMC1
Entrez Gene ID
375484
Gene Name
SUMO interacting motifs containing 1
Gene Aliases
C5orf25, OOMA1, PLEIAD
Chromosome Location
Chr. 5: 176238359 - 176345991 on Build GRCh38
UniGene ID
Hs.719847
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198567.5 NP_940969.3 1, 2 283
NR_131772.1 1, 2 283
GenBank mRNA BC066980.1 - 1, 2 276

View Full Product Details