Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015030.1 | NP_055845.1 | 6 | 843 | |
XM_005248090.4 | XP_005248147.1 | 6 | 856 | ||
XM_011513680.2 | XP_011511982.1 | 3 | 318 | ||
XM_011513681.2 | XP_011511983.1 | 3 | 318 | ||
XM_011513682.2 | XP_011511984.1 | 3 | 318 | ||
XM_011513683.2 | XP_011511985.1 | 5 | 692 | ||
XM_011513685.2 | XP_011511987.1 | 3 | 318 | ||
XM_017008046.1 | XP_016863535.1 | 7 | 1168 | ||
GenBank mRNA | AK131250.1 | - | 4 | 518 | |
AL833444.1 | - | 6 | 803 |