Gene Transcripts
- Gene Symbol
- RMDN3
- Entrez Gene ID
- 55177
- Gene Name
- regulator of microtubule dynamics 3
- Gene Aliases
- FAM82A2, FAM82C, RMD-3, RMD3, ptpip51
- Chromosome Location
- Chr. 15: 40735884 - 40755336 on Build GRCh38
- UniGene ID
- Hs.511067
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001304802.1 |
NP_001291731.1
|
11, 12 |
1501 |
|
NM_001323894.1 |
NP_001310823.1
|
11, 12 |
1415 |
|
NM_001323895.1 |
NP_001310824.1
|
11, 12 |
1632 |
|
NM_001323896.1 |
NP_001310825.1
|
11, 12 |
1493 |
|
NM_001323897.1 |
NP_001310826.1
|
11, 12 |
1609 |
|
NM_018145.2 |
NP_060615.1
|
11, 12 |
1531 |
|
XM_011521755.2 |
XP_011520057.1
|
8, 9 |
1777 |
|
GenBank mRNA |
AB000782.1 |
- |
10, 11 |
1270 |
|
AK001441.1 |
- |
11, 12 |
1455 |
|
AK025963.1 |
- |
8, 9 |
844 |
|
AK090731.1 |
- |
4, 5 |
1403 |
|
AK092058.1 |
- |
11, 12 |
1414 |
|
AK097286.1 |
- |
12, 13 |
1343 |
|
AK123192.1 |
- |
12, 13 |
1408 |
|
AK123282.1 |
- |
10, 11 |
2338 |
|
AY358793.1 |
- |
10, 11 |
1271 |
|
BC008970.2 |
- |
7, 8 |
728 |
|
BC063844.1 |
- |
11, 12 |
1419 |
6527521 |
View Full Product Details