Gene Transcripts

Gene Symbol
LRRC36
Entrez Gene ID
55282
Gene Name
leucine rich repeat containing 36
Gene Aliases
RORBP70, XLHSRF2
Chromosome Location
Chr. 16: 67326844 - 67385206 on Build GRCh38
UniGene ID
Hs.125139
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161575.1 NP_001155047.1 5 627
NM_018296.5 NP_060766.5 8 897
XM_005256025.2 XP_005256082.1 8 897
XM_005256027.2 XP_005256084.1 8 897
XM_005256028.1 XP_005256085.1 7 769
XM_011523200.1 XP_011521502.1 8 897
XM_011523201.1 XP_011521503.1 5 517
XM_011523202.2 XP_011521504.1 4 372
XM_011523204.1 XP_011521506.1 6 696
XM_011523205.1 XP_011521507.1 7 787
XM_011523206.2 XP_011521508.1 8 897
XM_017023400.1 XP_016878889.1 8 897
GenBank mRNA AK001866.1 - 5 676
AK292311.1 - 5 606
AK301766.1 - 5 581
AY313780.1 - 5 590
AY963267.1 - 8 897
BC026156.2 - 8 897 4828487

View Full Product Details