Gene Transcripts

Gene Symbol
ABCB10
Entrez Gene ID
23456
Gene Name
ATP binding cassette subfamily B member 10
Gene Aliases
EST20237, M-ABC2, MTABC2
Chromosome Location
Chr. 1: 229516582 - 229558987 on Build GRCh38
UniGene ID
Hs.17614
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012089.2 NP_036221.2 3 821
XM_011544135.2 XP_011542437.1 3 1592
GenBank mRNA AB013380.1 - 3 811
AF216833.1 - 3 821
AK172794.1 - 2 77
BC064930.1 - 3 816 6143235

View Full Product Details