Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018467.3 | NP_060937.1 | 3 | 236 | |
XM_017026975.1 | XP_016882464.1 | 4 | 946 | ||
XM_017026976.1 | XP_016882465.1 | 2 | 186 | ||
GenBank mRNA | AB097050.1 | - | 3 | 207 | |
AF220052.1 | - | 3 | 207 | ||
AK074683.1 | - | 3 | 192 | ||
BC006005.1 | - | 3 | 192 | 4279653 | |
BC008455.1 | - | 3 | 184 | 4247241 | |
BI758045.1 | - | 3 | 223 | ||
DA506853.1 | - | 2 | 186 |