Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001071775.2 | NP_001065243.1 | 3 | 1002 | |
GenBank mRNA | AK025522.1 | - | 3 | 875 | |
BC047655.1 | - | 1 | 513 | ||
BC062361.1 | - | 3 | 835 | ||
BC080523.1 | - | 3 | 851 | ||
BC125183.1 | - | 3 | 1005 | 40122127 |