Gene Transcripts

Gene Symbol
FAAP20
Entrez Gene ID
199990
Gene Name
Fanconi anemia core complex associated protein 20
Gene Aliases
C1orf86, FP7162
Chromosome Location
Chr. 1: 2184460 - 2212720 on Build GRCh38
UniGene ID
Hs.107101
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146310.1 NP_001139782.1 5 791
NM_001282670.1 NP_001269599.1 4 471
XM_011540914.2 XP_011539216.1 5 476
XM_011540921.2 XP_011539223.1 2 325
XM_017000553.1 XP_016856042.1 3 323
GenBank mRNA AK126870.1 - 4 471
AK127994.1 - 5 791

View Full Product Details