Gene Transcripts
- Gene Symbol
- DPY19L2P3
- Entrez Gene ID
- 442524
- Gene Name
- DPY19L2 pseudogene 3
- Gene Aliases
- Chromosome Location
- Chr. 7: 29685154 - 29688243 on Build GRCh38
- UniGene ID
- Hs.652669
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DQ287933.1 |
- |
9 |
682 |
|
- Gene Symbol
- LOC101928208
- Entrez Gene ID
- 101928208
- Gene Name
- probable C-mannosyltransferase DPY19L2
- Gene Aliases
- Chromosome Location
- Chr. 7: 29720476 - 29742396 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details