Gene Transcripts

Gene Symbol
KIF5C
Entrez Gene ID
3800
Gene Name
kinesin family member 5C
Gene Aliases
CDCBM2, KINN, NKHC, NKHC-2, NKHC2
Chromosome Location
Chr. 2: 148875223 - 149026759 on Build GRCh38
UniGene ID
Hs.435557
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004522.2 NP_004513.1 10, 11 1373
NR_111932.1 3, 4 395
XM_011511157.2 XP_011509459.1 3, 4 454
XM_017004062.1 XP_016859551.1 10, 11 1364
GenBank mRNA AB011103.1 - 10, 11 1346
BC002721.2 - 10, 11 1355
BC017298.2 - 10, 11 1328
BC025961.1 - 10, 11 1355
BC110287.1 - 3, 4 325 5165262
BC127772.1 - 7, 8 697
HY003338.1 - 3, 4 398

View Full Product Details