Gene Transcripts

Gene Symbol
MAST3
Entrez Gene ID
23031
Gene Name
microtubule associated serine/threonine kinase 3
Gene Aliases
Chromosome Location
Chr. 19: 18097764 - 18151692 on Build GRCh38
UniGene ID
Hs.466184
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015016.1 NP_055831.1 20 2182
XM_005259823.3 XP_005259880.1 20 2546
XM_005259824.2 XP_005259881.1 21 2232
XM_005259825.2 XP_005259882.1 21 2229
XM_005259826.3 XP_005259883.1 19 2342
XM_005259827.2 XP_005259884.1 19 2200
XM_006722698.2 XP_006722761.1 20 2224
XM_006722699.2 XP_006722762.2 20 2533
XM_006722700.3 XP_006722763.1 19 2342
XM_011527823.1 XP_011526125.1 22 2322
XM_017026506.1 XP_016881995.1 20 2530
XM_017026507.1 XP_016881996.1 19 2512
XM_017026508.1 XP_016881997.1 19 2510
XM_017026509.1 XP_016881998.1 20 2533
XM_017026510.1 XP_016881999.1 19 2513
XM_017026511.1 XP_016882000.1 21 2620
XM_017026512.1 XP_016882001.1 20 2211
XM_017026513.1 XP_016882002.1 20 2208
XM_017026514.1 XP_016882003.1 21 2215
XM_017026515.1 XP_016882004.1 20 2221
XM_017026516.1 XP_016882005.1 20 2530
GenBank mRNA AB011133.1 - 20 2180

View Full Product Details