Gene Transcripts

Gene Symbol
ZC4H2
Entrez Gene ID
55906
Gene Name
zinc finger C4H2-type containing
Gene Aliases
HCA127, KIAA1166, WRWF, WWS
Chromosome Location
Chr. X: 64915802 - 65034744 on Build GRCh38
UniGene ID
Hs.28249
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001178032.2 NP_001171503.1 5 1894
NM_001178033.2 NP_001171504.1 4 1569
NM_001243804.1 NP_001230733.1 5 1639
NM_018684.3 NP_061154.1 5 1732
NR_045044.1 5 1987
GenBank mRNA AB032992.1 - 3 3753
AF270491.1 - 5 1664
AK001547.1 - 2 1161
AK022807.1 - 5 1863
AK022918.1 - 5 1626
AK123763.1 - 6 2125
BC004411.1 - 5 1683 3639978

View Full Product Details