Gene Transcripts

Gene Symbol
SYCN
Entrez Gene ID
342898
Gene Name
syncollin
Gene Aliases
INSSA1, SYL
Chromosome Location
Chr. 19: 39202922 - 39204266 on Build GRCh38
UniGene ID
Hs.124123
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080468.2 NP_001073937.1 1 140
GenBank mRNA AK130951.1 - 1 151
BC121075.2 - 1 144 40119665
BC121076.2 - 1 144 40119666

View Full Product Details