Gene Transcripts

Gene Symbol
NLGN3
Entrez Gene ID
54413
Gene Name
neuroligin 3
Gene Aliases
HNL3
Chromosome Location
Chr. X: 71144389 - 71175307 on Build GRCh38
UniGene ID
Hs.438877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166660.1 NP_001160132.1 3 847
NM_001321276.1 NP_001308205.1 4 394
NM_018977.3 NP_061850.2 4 907
NM_181303.1 NP_851820.1 5 967
XM_006724662.3 XP_006724725.2 6 782
XM_006724663.3 XP_006724726.1 6 974
XM_011530974.2 XP_011529276.1 5 896
XM_017029597.1 XP_016885086.1 4 769
GenBank mRNA AB040913.1 - 2 137
AF217411.1 - 4 868
AF217412.1 - 4 679
AK296217.1 - 2 202
AK299567.1 - 2 520
AK299669.1 - 4 378
AK314699.1 - 3 684
BC028738.1 - 3 842
BC051715.1 - 4 837 5258980
BX641059.1 - 4 390
GQ489207.1 - 4 633
KJ534889.1 - 3 573
KJ534890.1 - 2 513
KJ534891.1 - 4 633
KJ534977.1 - 2 513
KJ535034.1 - 3 573
KJ535040.1 - 2 513
KU178651.1 - 3 573

View Full Product Details