Gene Transcripts
- Gene Symbol
- CCDC93
- Entrez Gene ID
- 54520
- Gene Name
- coiled-coil domain containing 93
- Gene Aliases
- Chromosome Location
- Chr. 2: 117915478 - 118014163 on Build GRCh38
- UniGene ID
- Hs.107845
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019044.4 |
NP_061917.3
|
5, 6 |
638 |
|
XM_006712600.2 |
XP_006712663.1
|
5, 6 |
638 |
|
XM_011511359.1 |
XP_011509661.1
|
5, 6 |
638 |
|
XM_011511361.1 |
XP_011509663.1
|
4, 5 |
392 |
|
XM_011511362.1 |
XP_011509664.1
|
4, 5 |
506 |
|
XM_011511363.2 |
XP_011509665.1
|
5, 6 |
638 |
|
GenBank mRNA |
AB209993.1 |
- |
5, 6 |
557 |
|
AK025611.1 |
- |
4, 5 |
406 |
|
AK057926.1 |
- |
5, 6 |
499 |
|
AK290722.1 |
- |
5, 6 |
545 |
|
AK309040.1 |
- |
5, 6 |
545 |
|
BC028609.1 |
- |
5, 6 |
611 |
4826022 |
BC093018.1 |
- |
5, 6 |
528 |
30337330 |
View Full Product Details