Gene Transcripts

Gene Symbol
WDR44
Entrez Gene ID
54521
Gene Name
WD repeat domain 44
Gene Aliases
RAB11BP, RPH11
Chromosome Location
Chr. X: 118345997 - 118449961 on Build GRCh38
UniGene ID
Hs.98510
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184965.1 NP_001171894.1 10 1916
NM_001184966.1 NP_001171895.1 9 1841
NM_019045.4 NP_061918.3 10 1916
XM_011531353.2 XP_011529655.1 9 1917
XM_017029600.1 XP_016885089.1 9 1849
GenBank mRNA AK001978.1 - 4 384
AK092077.1 - 7 920
AK127556.1 - 10 1827
AK299711.1 - 9 1777
AK304432.1 - 10 1443
AL833605.1 - 10 1881
AL834210.1 - 7 1183
BC028697.2 - 10 1852 4837674
BX538087.1 - 10 1858
BX648150.1 - 10 1855

View Full Product Details