Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033390.1 | NP_203748.1 | 6 | 2135 | |
XM_005271715.3 | XP_005271772.1 | 6 | 2516 | ||
XM_011543055.2 | XP_011541357.1 | 6 | 2248 | ||
XM_017018477.1 | XP_016873966.1 | 6 | 6686 | ||
GenBank mRNA | AB051513.1 | - | 4 | 1308 | |
AB096241.1 | - | 5 | 2872 | ||
AK295437.1 | - | 6 | 2136 | ||
AK299100.1 | - | 6 | 2168 | ||
AL832893.1 | - | 3 | 988 |