Gene Transcripts

Gene Symbol
RPS21
Entrez Gene ID
6227
Gene Name
ribosomal protein S21
Gene Aliases
HLDF, S21
Chromosome Location
Chr. 20: 62387065 - 62388520 on Build GRCh38
UniGene ID
Hs.190968
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024.3 NP_001015.1 4 228
GenBank mRNA AV758927.1 - 189
BC018140.1 - 3 165 3903320
BC021990.1 - 4 193 4066330
BM840443.1 - 4 228
CF124568.1 - 4 269
CR542132.1 - 3 138
L04483.1 - 4 176
X79563.2 - 4 174

View Full Product Details