Gene Transcripts

Gene Symbol
LCN15
Entrez Gene ID
389812
Gene Name
lipocalin 15
Gene Aliases
PRO6093, UNQ2541
Chromosome Location
Chr. 9: 136759634 - 136764513 on Build GRCh38
UniGene ID
Hs.413902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203347.1 NP_976222.1 1 108
XM_011518672.1 XP_011516974.1 1 80
GenBank mRNA AY358618.1 - 1 108
BC035124.1 - 1 1854 5263500
BC093893.1 - 1 103 7939738
BC093895.1 - 1 103 7939740

View Full Product Details