Gene Transcripts

Gene Symbol
BLID
Entrez Gene ID
414899
Gene Name
BH3-like motif containing, cell death inducer
Gene Aliases
BRCC2
Chromosome Location
Chr. 11: 122115354 - 122116215 on Build GRCh38
UniGene ID
Hs.686109
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001786.2 NP_001001786.2 1 535
GenBank mRNA AF303179.1 - 1 535
BC130361.1 - 1 413 40146392
BC130363.1 - 1 413 40146394

View Full Product Details