Gene Transcripts

Gene Symbol
NHLRC4
Entrez Gene ID
283948
Gene Name
NHL repeat containing 4
Gene Aliases
Chromosome Location
Chr. 16: 566995 - 569495 on Build GRCh38
UniGene ID
Hs.664267
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301159.1 NP_001288088.1 2 993
NM_176677.2 NP_788850.1 2 968
GenBank mRNA AK093527.1 - 2 951
BC032015.1 - 2 931
BC048340.1 - 2 930
BC113527.1 - 1 853 8322579
BC113529.1 - 1 853 8322581

View Full Product Details