Gene Transcripts

Gene Symbol
UGGT2
Entrez Gene ID
55757
Gene Name
UDP-glucose glycoprotein glucosyltransferase 2
Gene Aliases
HUGT2, UGCGL2, UGT2
Chromosome Location
Chr. 13: 95801580 - 96053482 on Build GRCh38
UniGene ID
Hs.193226
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020121.3 NP_064506.3 1, 2 331
XM_011521094.1 XP_011519396.1 1, 2 286
XM_011521095.1 XP_011519397.1 1, 2 286
XM_011521096.1 XP_011519398.1 1, 2 286
XM_011521097.1 XP_011519399.1 1, 2 286
XM_011521100.2 XP_011519402.1 1, 2 286
XM_017020662.1 XP_016876151.1 1, 2 286
XM_017020663.1 XP_016876152.1 1, 2 250
GenBank mRNA AF227906.2 - 1, 2 232
BC017759.1 - 1, 2 235
BC032302.1 - 1, 2 331
BC067807.1 - 1, 2 242
BC111021.1 - 1, 2 249
BC125233.1 - 1, 2 187 40124676
BC125234.1 - 1, 2 187

View Full Product Details